Citric acid ≥99.5% for electrophoresis
Supplier: Thermo Scientific Chemicals
|
Warning
|
J64322-22
J64322-36
J64322-A3
CAAAJ64322-22EA
36.97
CAD
CAAAJ64322-22
CAAAJ64322-36
CAAAJ64322-A3
Citric acid ≥99.5% for electrophoresis
Citric acid
Powder
Purity: 99.5+%
CAS No.: 77-92-9
Molecular Formula: C6H8O7
Formula Weight: 192.13
Physical Form: Powder
Density: 1.665
Appearance: White
Odor: Odorless
pH: 1,6
Melting Point: 153-154°
Beilstein Reference: 782061
Merck Reference: 14,2326
MDL No.: MFCD00011669
Formula:
HOC(COOH)(CH₂COOH)₂ Boiling Pt: 310 °C (1013 hPa) Melting Pt: 153…154 °C Density: 1.665 Storage Temperature: Ambient |
MDL Number:
MFCD00011669 CAS Number: 77-92-9 EINECS: 201-069-1 REACH: 01-2119457026-42-0000 Merck Index: 14,2326 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...