Bromocresol green sodium salt ACS
Supplier: Thermo Scientific Chemicals
Synonyms:
Bromocresol green water soluble, Sodium 2,6-dibromo-4-(3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-2,2-dioxido-3H-benzo[d][1,2]oxathiol-3-yl)-3-methylphenolate
J64284-06
J64284-14
CAAAJ64284-06EA
67.15
CAD
CAAAJ64284-06
CAAAJ64284-14
Bromocresol green sodium salt ACS
Bromocresol green sodium salt
Crystalline powder
CAS No.: 62625-32-5
Molecular Formula: C21H13Br4NaO5S
Formula Weight: 720.02
Physical Form: Crystalline powder
Appearance: Dark brown to dark red
Melting Point: 230° dec.
MDL No.: MFCD00148898
Formula:
C₂₁H₁₃Br₄NaO₅S MW: 720 g/mol Melting Pt: 225 (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00148898 CAS Number: 62625-32-5 EINECS: 263-657-4 Merck Index: 14,1386 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...