D-(+)-Sucrose 99%, Ultrapure
Supplier: Thermo Scientific Chemicals
J64270-22
J64270-36
J64270-A1
CAAAJ64270-22EA
36.89
CAD
CAAAJ64270-22
CAAAJ64270-36
CAAAJ64270-A1
D-(+)-Sucrose 99%, Ultrapure
D(+)-Sucrose
Crystalline
Purity: 99%
CAS No.: 57-50-1
Molecular Formula: C12H22O11
Formula Weight: 342.30
Physical Form: Crystalline
Density: 1.58
Appearance: White
Odor: Odorless
Melting Point: 185-187°
Beilstein Reference: 90825
Merck Reference: 14,8881
MDL No.: MFCD00006626
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 185…187 °C Density: 1.58 Storage Temperature: Ambient |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 Merck Index: 14,8881 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...