2-Amino-2'-deoxyadenosine 99%
Supplier: Thermo Scientific Chemicals
J64266-14
J64266-03
J64266-06
CAAAJ64266-14EA
225.94
CAD
CAAAJ64266-14
CAAAJ64266-03
CAAAJ64266-06
2-Amino-2'-deoxyadenosine 99%
2-Amino-2'-deoxyadenosine
Powder
Purity: 99%
CAS No.: 4546-70-7
Molecular Formula: C10H14N6O3
Formula Weight: 266.26
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: White to off-white
MDL No.: MFCD00047240
Formula:
C₁₀H₁₄N₆O₃ Storage Temperature: Ambient |
MDL Number:
MFCD00047240 CAS Number: 4546-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...