Sodium dodecyl sulfate (SDS) 97% for electrophoresis

Supplier: Thermo Scientific Chemicals
Danger

J64241-14 J64241-22 J64241-36
CAAAJ64241-14EA 75.52 CAD
CAAAJ64241-14 CAAAJ64241-22 CAAAJ64241-36
Sodium dodecyl sulfate (SDS) 97% for electrophoresis
Sodium dodecyl sulphate (SDS)

Powder


Purity: 98% CAS No.: 151-21-3 Molecular Formula: C12H25NaO4S Formula Weight: 288.38 Physical Form: Powder Appearance: White Melting Point: ca 206° Solubility: Soluble in water Beilstein Reference: 3599286 Merck Reference: 14,8636 UN No.: 2926 MDL No.: MFCD00036175

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: ∼ 206 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: ≥ 100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1,III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR