Salubrinal
Supplier: Thermo Scientific Chemicals
J64192-LB0
J64192-MA
CAAAJ64192-LB0EA
188.02
CAD
CAAAJ64192-LB0
CAAAJ64192-MA
Salubrinal
Salubrinal
Powder
CAS No.: 405060-95-9
Molecular Formula: C21H17Cl3N4OS
Formula Weight: 479.81
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Pale brown
MDL No.: MFCD00548612
Formula:
C₂₁H₁₇Cl₃N₄OS Storage Temperature: Ambient |
MDL Number:
MFCD00548612 CAS Number: 405060-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...