Cytidine-5'-diphosphocholine sodium salt (CDP choline), hydrate

Supplier: Thermo Scientific Chemicals

J64161-09 J64161-14 J64161-18
CAAAJ64161-09EA 108.23 CAD
CAAAJ64161-09 CAAAJ64161-14 CAAAJ64161-18
Cytidine-5'-diphosphocholine sodium salt (CDP choline), hydrate
Cytidine-5'-diphosphocholine sodium salt (CDP choline)

Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor


Powder


Applications: Intermediate in the synthesis of phosphatidylcholine; also a phospholipase inhibitor

Formula: C₁₄H₂₆N₄NaO₁₁P₂
Storage Temperature: Ambient
MDL Number: MFCD00037486
CAS Number: 33818-15-4
EINECS: 251-689-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR