Cathepsin K Substrate (fluorogenic)

Supplier: Thermo Scientific Chemicals

J64082-MA J64082-MB
CAAAJ64082-MAEA 240.16 CAD
CAAAJ64082-MA CAAAJ64082-MB
Cathepsin K Substrate (fluorogenic)
Cathepsin K substrate (fluorogenic)

A sensitive fluorogenic substrate for cathepsin K


Molecular Formula: C30H38N6O6 Formula Weight: 578.65 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: White Solubility: Soluble in DMSO MDL No.: MFCD07780911


Applications: A sensitive fluorogenic substrate for cathepsin K

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR