Amoxapine 98%

Supplier: Thermo Scientific Chemicals
Warning

J64066-03 J64066-06 J64066-MD
CAAAJ64066-03EA 358.66 CAD
CAAAJ64066-03 CAAAJ64066-06 CAAAJ64066-MD
Amoxapine 98%
Amoxapine

A tricyclic norepinephrine uptake inhibitor


Powder


Applications: A tricyclic norepinephrine uptake inhibitor

Formula: C₁₇H₁₆ClN₃O
Storage Temperature: Ambient
MDL Number: MFCD00069210
CAS Number: 14028-44-5
EINECS: 237-867-1
Merck Index: 14,576

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR