4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid

Supplier: Thermo Scientific Chemicals

Synonyms: Tamibarotene

J64028-MA J64028-MF
CAAAJ64028-MAEA 244.9 CAD
CAAAJ64028-MA CAAAJ64028-MF
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid

A retinoic acid receptor _x001A_ (RAR_x001A_) agonist


Solid


Applications: A retinoic acid receptor (RAR) agonist

Formula: C₂₂H₂₅NO₃
MW: 351.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00866188
CAS Number: 94497-51-5
Merck Index: 14,9047

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR