Phenol 99% unstabilised, Ultrapure

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Phenylalcohol, Carbolic acid, Hydroxybenzene

J64011-22 J64011-36
CAAAJ64011-22EA 95.8 CAD
CAAAJ64011-22 CAAAJ64011-36
Phenol 99% unstabilised, Ultrapure
Phenol

Crystalline


Purity: 99% CAS No.: 108-95-2 Molecular Formula: C6H5OH Formula Weight: 94.11 Flash Point: 79°(175°F) Physical Form: Crystalline Density: 1.071 Sensitivity: Air Sensitive Appearance: White Odor: Sweet Melting Point: 40-43° Boiling Point: 181° Beilstein Reference: 969616 Merck Reference: 14,7241 UN No.: 1671 MDL No.: MFCD00002143

Formula: C₆H₅OH
Boiling Pt: 181 °C
Melting Pt: 40…43 °C
Density: 1.071
Flash Pt: 79 °C (175 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002143
CAS Number: 108-95-2
EINECS: 203-632-7
UN: 1671
ADR: 6.1,II
REACH: 01-2119471329-32
Merck Index: 14,7241

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR