2-Mercaptoethanesulfonic acid sodium salt (mesna) 96%

Supplier: Thermo Scientific Chemicals
Warning

J63989-09 J63989-14
CAAAJ63989-09EA 332.97 CAD
CAAAJ63989-09 CAAAJ63989-14
2-Mercaptoethanesulfonic acid sodium salt (mesna) 96%
2-Mercaptoethanesulphonic acid sodium salt (Mesna)

Water-soluble disulfide reducing agent. Cofactor in methyl transfer reactions.


Powder


Water-soluble disulfide reducing agent. Cofactor in methyl transfer reactions

Formula: C₂H₅NaO₃S₂
Storage Temperature: Ambient
MDL Number: MFCD00007535
CAS Number: 19767-45-4
EINECS: 243-285-9
Merck Index: 14,5909

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR