Ticlopidine Hydrochloride
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Triclopidine hydrochloride, Ticlopidine HCl, 5-(2-Chlorobenzyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine Hydrochloride
J63971-06
J63971-14
CAAAJ63971-06EA
165.16
CAD
CAAAJ63971-06
CAAAJ63971-14
Ticlopidine Hydrochloride
Ticlopidine hydrochloride
Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Powder
Applications: Platelet aggregation inhibitor capable of inducing apoptosis in various cancer cell lines
Formula:
C₁₄H₁₄ClNS MW: 300.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00079606 CAS Number: 53885-35-1 EINECS: 258-837-4 Merck Index: 14,9429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...