(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Nα-p-Tosyl-DL-lysine chloromethyl ketone hydrochloride, TLCK, Nalpha-p-Tosyl-DL-lysine chloromethyl ketone hydrochloride
J63959-MC
J63959-MF
CAAAJ63959-MCEA
410.94
CAD
CAAAJ63959-MC
CAAAJ63959-MD
CAAAJ63959-MF
(3S)-1-Chloro-3-tosylamido-7-amino-2-heptanone hydrochloride 98%
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride
Inhibitor of papain and trypsin
Powder
Applications: Inhibitor of papain and trypsin
Formula:
C₁₄H₂₂Cl₂N₂O₃S MW: 369.31 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00012513 CAS Number: 4238-41-9 EINECS: 224-199-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...