Ethyl violet
Supplier: Thermo Scientific Chemicals
Synonyms:
Basic Violet 4, N-(4-(Bis(4-(diethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-ethylethanaminium chloride
J63951-14
J63951-22
CAAAJ63951-14EA
75.05
CAD
CAAAJ63951-14
CAAAJ63951-22
Ethyl violet
Ethyl violet
Powder
CAS No.: 2390-59-2
Molecular Formula: C31H42ClN3
Formula Weight: 492.15
Physical Form: Powder
Appearance: Olive green
Beilstein Reference: 4117566
MDL No.: MFCD00011765
Formula:
C₃₁H₄₂ClN₃ MW: 560.29 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00011765 CAS Number: 2390-59-2 EINECS: 219-231-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...