Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63950-AK J63950-AP
CAAAJ63950-AKEA 175.14 CAD
CAAAJ63950-AK CAAAJ63950-AP
Citrate, 0.5 M Buffer Solution, pH 6.0, Thermo Scientific
Buffers Biological Buffers

Liquid


CAS No.: 6132-04-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00150031

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR