L-Phenylalanine Cell Culture

Supplier: Thermo Scientific Chemicals

Synonyms: H-L-Phe-OH, Phenylalanine, H-Phe-OH, L-α-Phenylalanine, (S)-(-)-Phenylalanine, 3-Phenyl-L-alanine, 3-Phenyl-L(-)-alanine

J63925-22 J63925-30 J63925-36
CAAAJ63925-22EA 287.64 CAD
CAAAJ63925-22 CAAAJ63925-30 CAAAJ63925-36
L-Phenylalanine Cell Culture
L-α-Phenylalanine

Powder


CAS No.: 63-91-2 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Physical Form: Powder Appearance: White to off-white Solubility: Soluble in water. Aqueous solubility increases with warming. Also soluble in methanol, ethanol or 1M NH4OH Beilstein Reference: 1910408 Merck Reference: 14,7271 MDL No.: MFCD00064227

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
Boiling Pt: 313
Melting Pt: 173
Storage Temperature: Ambient
MDL Number: MFCD00064227
CAS Number: 63-91-2
EINECS: 200-568-1
Merck Index: 14,07271

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR