Isradipine ≥98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Isopropyl Methyl 4-(4-Benzofurazanyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylate, 3-Isopropyl 5-methyl 4-(benzo[c][1,2,5]oxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 4-(4-Benzofurazanyl)-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic acid Isopropyl Methyl Ester, Isopropyl Methyl 4-(2,1,3-Benzoxadiazol-4-yl)-1,4-dihydro-2,6-dim
L-type calcium channel blocker
Crystalline powder
Applications: L-type calcium channel blocker
Formula:
C₁₉H₂₁N₃O₅ MW: 371.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00153820 CAS Number: 75695-93-1 Merck Index: 14,5240 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...