Dizocilpine maleate ≥99%

Supplier: Thermo Scientific Chemicals

J63917-MA J63917-MF
CAAAJ63917-MAEA 241.53 CAD
CAAAJ63917-MA CAAAJ63917-MF
Dizocilpine maleate ≥99%
Dizocilpine maleate

NMDA receptor antagonist that prevents calcium flux


Crystalline powder


Applications: NMDA receptor antagonist that prevents calcium flux

Formula: C₁₆H₁₅N·C₄H₄O₄
Storage Temperature: Ambient
MDL Number: MFCD00082465
CAS Number: 77086-22-7
EINECS: 278-614-5
Merck Index: 14,3386

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR