(Hydroxy-2-naphthylmethyl)phosphonic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63913-LB0
J63913-MB
CAAAJ63913-LB0EA
310.83
CAD
CAAAJ63913-LB0
CAAAJ63913-MB
(Hydroxy-2-naphthylmethyl)phosphonic acid 98%
(Hydroxy-2-naphthylmethyl)phosphonic acid
A selective insulin receptor tyrosine kinase inhibitor
Purity: 98%
CAS No.: 120943-99-9
Molecular Formula: C11H11O4P
Formula Weight: 238.18
Storage Temperature: -30°C to -10°C
Physical Form: Solid
MDL No.: MFCD00674174
A selective insulin receptor tyrosine kinase inhibitor
Formula:
C₁₁H₁₁O₄P Storage Temperature: Refrigerator |
MDL Number:
MFCD00674174 CAS Number: 120943-99-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...