Penicillin G potassium salt

Supplier: Thermo Scientific Chemicals
Danger

J63901-14 J63901-22
CAAAJ63901-14EA 101.96 CAD
CAAAJ63901-14 CAAAJ63901-22
Penicillin G potassium salt
Penicillin G potassium salt

Inhibits bacterial cell wall synthesis


Powder


Inhibits bacterial cell wall synthesis

Formula: C₁₆H₁₇KN₂O₄S
Melting Pt: 214…217 °C
Density: 1.359 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00036193
CAS Number: 113-98-4
EINECS: 204-038-0
Merck Index: 14,7094

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR