Clotrimazole

Supplier: Thermo Scientific Chemicals
Warning

J63895-06 J63895-14
CAAAJ63895-06EA 186.3 CAD
CAAAJ63895-06 CAAAJ63895-14
Clotrimazole
Clotrimazole

Specific inhibitor of calcium-activated potassium channels


Powder


Applications: Specific inhibitor of calcium-activated potassium channels

Formula: C₂₂H₁₇ClN₂
Melting Pt: 141…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00057220
CAS Number: 23593-75-1
EINECS: 245-764-8
Merck Index: 14,2417

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR