Sodium Citrate, 0.5 M Buffer Solution, pH 6.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63888-AK
J63888-AP
CAAAJ63888-AKEA
150.07
CAD
CAAAJ63888-AK
CAAAJ63888-AP
Sodium Citrate, 0.5 M Buffer Solution, pH 6.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00012462
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...