Imidazole-buffered saline (5X), Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63887-AK J63887-AP
CAAAJ63887-AKEA 100.82 CAD
CAAAJ63887-AK CAAAJ63887-AP
Imidazole-buffered saline (5X), Thermo Scientific
Buffers Electrophoresis Buffers Washing Buffers

Liquid


CAS No.: 288-32-4
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00005183

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR