2'-Deoxyadenosine 99%

Supplier: Thermo Scientific Chemicals
Warning

J63886-18 J63886-22
CAAAJ63886-18EA 527.99 CAD
CAAAJ63886-18 CAAAJ63886-22
2'-Deoxyadenosine 99%
2'-Deoxyadenosine

Powder


Purity: 99% CAS No.: 958-09-8 Molecular Formula: C10H13N5O3 Formula Weight: 251.24 Physical Form: Powder Appearance: White Solubility: Soluble in water at 50mg/ml Optical Rotation: -25° (c=0.5 in water) MDL No.: MFCD00056003

Formula: C₁₀H₁₃N₅O₃
Storage Temperature: Ambient
MDL Number: MFCD00056003
CAS Number: 958-09-8
EINECS: 213-488-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR