Succinate, 0.2 M Buffer Solution, pH 4.5, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63881-AK
J63881-AP
CAAAJ63881-AKEA
170.01
CAD
CAAAJ63881-AK
CAAAJ63881-AP
Succinate, 0.2 M Buffer Solution, pH 4.5, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 150-90-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00002789
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...