HEPPSO, 0.2 M Buffer Solution, pH 7.0, Thermo Scientific
Supplier: Thermo Scientific Chemicals
J63877-AE
J63877-AK
CAAAJ63877-AEEA
169.96
CAD
CAAAJ63877-AE
CAAAJ63877-AK
HEPPSO, 0.2 M Buffer Solution, pH 7.0, Thermo Scientific
Buffers
Biological Buffers
Liquid
CAS No.: 68399-78-0
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00038351
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...