G418 disulfate 50 mg/ml in solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: G418 disulfate, G-418 sulfate, G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate, G-418 disulfate

J63871-AB J63871-AD
CAAAJ63871-ABEA 275.43 CAD
CAAAJ63871-AB CAAAJ63871-AD
G418 disulfate 50 mg/ml in solution
G418 disulphate

Aminoglycoside that is toxic to bacteria, yeast, protozoans and helminths


Clear liquid

Formula: C₂₀H₄₄N₄O₁₈S₂
MW: 692.72 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR