Agarose Gel Loading Dye 6X for Electrophoresis, Glycerol Based, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63869-AC J63869-AD
CAAAJ63869-ACEA 121.41 CAD
CAAAJ63869-AC CAAAJ63869-AD
Agarose Gel Loading Dye 6X for Electrophoresis, Glycerol Based, Thermo Scientific
Stains Electrophoresis Stains

Agarose gel loading dyes (6X), alkaline are used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.


  • Liquid, Fully miscible in water
  • Store at Cold Temperature


Contains 0.25% bromophenol blue, 0.25% xylene cyanol FF and 40% glycerol.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR