Indapamide

Supplier: Thermo Scientific Chemicals

J63846-03 J63846-06
CAAAJ63846-03EA 111.25 CAD
CAAAJ63846-03 CAAAJ63846-06
Indapamide
Indapamide

Used as an antihypertensive. Diuretic


Powder


Applications: Used as an antihypertensive. Diuretic

Formula: C₁₆H₁₆ClN₃O₃S
Melting Pt: 160…162 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079375
CAS Number: 26807-65-8
EINECS: 248-012-7
Merck Index: 14,4935

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR