2,2'-O-Cyclocytidine Hydrochloride ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Ancitabine Hydrochloride, Cyclocytidine hydrochloride, (2R,3R,3aS,9aR)-2-(Hydroxymethyl)-6-imino-3,3a,6,9a-tetrahydro-2H-furo[2',3':4,5]oxazolo[3,2-a]pyrimidin-3-ol hydrochloride, 2,2'-Anhydro-(1-beta-D-arabinofuranosyl)cytosine Hydrochloride

J63845-06 J63845-03
CAAAJ63845-06EA 414.66 CAD
CAAAJ63845-06 CAAAJ63845-03
2,2'-O-Cyclocytidine Hydrochloride ≥98%
2,2'-O-Cyclocytidine hydrochloride

Anti-tumor agent


Powder


Applications: Anti-tumor agent

Formula: C₉H₁₁N₃O₄
MW: 261.66 g/mol
Melting Pt: 248…250 °C
Storage Temperature: Ambient
MDL Number: MFCD00012636
CAS Number: 10212-25-6
EINECS: 233-515-6
Merck Index: 14,629

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR