Dihydroergotamine mesylate

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dihydroergotamine methanesulfonate, Dihydroergotamine methanesulphonate

J63840-03 J63840-MC
CAAAJ63840-03EA 1469.72 CAD
CAAAJ63840-03 CAAAJ63840-MC
Dihydroergotamine mesylate
Dihydroergotamine mesylate

Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors.


Powder


Agonist at vascular serotonin receptors; partial agonist at alpha-adrenergic and dopamine D2 receptors

Formula: C₃₄H₄₁N₅O₈S
Melting Pt: 230…235 °C
Storage Temperature: Ambient
MDL Number: MFCD00058615
CAS Number: 6190-39-2
EINECS: 228-235-6
Merck Index: 14,3174

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR