Proadifen hydrochloride

Supplier: Thermo Scientific Chemicals

J63833-03 J63833-ME
CAAAJ63833-03EA 719.88 CAD
CAAAJ63833-03 CAAAJ63833-ME
Proadifen hydrochloride
Proadifen hydrochloride

Inhibits cytochrome P-450, blocks glibenclamide-sensitive potassium channels. Also inhibits neuronal nitric oxide synthase


Powder


Applications: Inhibits cytochrome P-450, blocks glibenclamide-sensitive potassium channels. Also inhibits neuronal nitric oxide synthase

Formula: C₂₃H₃₁NO₂·HCl
Melting Pt: 122…123 °C
Storage Temperature: Ambient
MDL Number: MFCD00055151
CAS Number: 62-68-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR