3,5-Dimethyl-1-adamantanamine hydrochloride

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Dimethyl-5-aminoadamantane hydrochloride1-Amino-3,5-dimethyladamantane HydrochlorideMemantine Hydrochloride

J63830-MF J63830-MF
CAAAJ63830-MFEA 257.18 CAD
AAJ63830-MF CAAAJ63830-MF
3,5-Dimethyl-1-adamantanamine hydrochloride
3,5-Dimethyl-1-adamantanamine hydrochloride

An NMDA receptor antagonist; stimulates dopamine release


Powder

Formula: C₁₂H₂₁N
MW: 215.77 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00214336
CAS Number: 41100-52-1
EINECS: 255-219-6
Merck Index: 14,5829

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR