Harmol hydrochloride dihydrate 98%
Supplier: Thermo Scientific Chemicals
J63827-03
J63827-06
CAAAJ63827-03EA
176.03
CAD
CAAAJ63827-03
CAAAJ63827-06
Harmol hydrochloride dihydrate 98%
Harmol hydrochloride dihydrate
Induces apoptosis by Caspase-8 activation
Powder
Applications: Induces apoptosis by Caspase-8 activation
Formula:
C₁₂H₁₀N₂O·HCl·2H₂O MW: 270.72 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150058 CAS Number: 149022-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...