Sodium Phosphate, 0.2 M Buffer Solution, pH 7.2, Thermo Scientific

Supplier: Thermo Scientific Chemicals

J63816-AP J63816-K2
CAAAJ63816-APEA 150.07 CAD
CAAAJ63816-AP CAAAJ63816-K2
Sodium Phosphate, 0.2 M Buffer Solution, pH 7.2, Thermo Scientific
Buffers Biological Buffers

Liquid


Physical Form: Liquid
MDL No.: MFCD00146206

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR