Ampicillin sodium salt

Supplier: Thermo Scientific Chemicals
Danger

J63807-06 J63807-09
CAAAJ63807-03EA 56.88 CAD
CAAAJ63807-03 CAAAJ63807-06 CAAAJ63807-09
Ampicillin sodium salt
Ampicillin sodium salt

Powder


CAS No.: 69-52-3 Molecular Formula: C16H18N3NaO4S Formula Weight: 371.39 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Melting Point: 215° Solubility: Freely soluble in water. Sparingly soluble in acetone Beilstein Reference: 4119211 Merck Reference: 14,586 MDL No.: MFCD00064313

Formula: C₁₆H₁₈N₃NaO₄S
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1
Merck Index: 14,586

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR