Doxycycline monohydrate

Supplier: Thermo Scientific Chemicals
Warning

J63805-06 J63805-14
CAAAJ63805-06EA 100.38 CAD
CAAAJ63805-06 CAAAJ63805-14
Doxycycline monohydrate
Doxycycline monohydrate

An antimicrobial. Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases


Powder


Applications: An antimicrobial. Inhibits angiogenesis and reduces lung metastases by inhibiting matrix metalloproteinases

Formula: C₂₂H₂₄N₂O₈·H₂O
MW: 462.46 g/mol (444.44 anhy)
Melting Pt: 167…168 °C
Storage Temperature: Ambient
MDL Number: MFCD02682958
CAS Number: 17086-28-1
Merck Index: 14,3440

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR