Yeast Hexokinase
Supplier: Thermo Scientific Chemicals
J63795-4I J63795-JPL
CAAAJ63795-4IEA 332.84 CAD
CAAAJ63795-4I CAAAJ63795-JPL
Yeast Hexokinase
Enzymes
Catalyzes the phosphorylation of D-hexose sugars at the C6 position utilizing ATP as a phosphate source. Used to determine glucose, fructose, mannose and ATP.
  • Hygroscopic