Dextran, M.W. ~ 150,000

Supplier: Thermo Scientific Chemicals

J63789-09 J63789-18 J63789-22 J63789-36 J63789-A1
CAAAJ63789-09EA 97.04 CAD
CAAAJ63789-09 CAAAJ63789-18 CAAAJ63789-22 CAAAJ63789-36 CAAAJ63789-A1
Dextran, M.W. ~ 150,000
Dextran

Powder


CAS No.: 9004-54-0 Molecular Formula: (C6H16O5)n Physical Form: Powder Appearance: White Merck Reference: 14,2948 MDL No.: MFCD00130935

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0
EINECS: 232-677-5
Merck Index: 14,2948

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR