Methyl caffeate

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4-Dihydroxycinnamic acid methyl ester, Methyl 3,4-dihydroxycinnamate, Caffeic acid methyl ester, Methyl 3-(3,4-dihydroxyphenyl)acrylate

J63786-03 J63786-MD
CAAAJ63786-03EA 709.77 CAD
CAAAJ63786-03 CAAAJ63786-MD
Methyl caffeate
Methyl caffeate

Inhibitor of protein tyrosine kinase


Powder


Applications: Inhibitor of protein tyrosine kinase

Formula: C₁₀H₁₀O₄
MW: 194.19 g/mol
Melting Pt: 158…161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00210468
CAS Number: 3843-74-1
Merck Index: 14,1635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR