DEAE-Dextran, powder
Supplier: Thermo Scientific Chemicals
J63781-14
J63781-22
CAAAJ63781-14EA
156.53
CAD
CAAAJ63781-14
CAAAJ63781-22
DEAE-Dextran, powder
DEAE-Dextran
DEAE-Dextran is used in the development of sustained release protein delivery and nucleic acid transfection systems.
It has been speculated that DEAE-Dextran may cause a stimulation of the helper T-cell function in antibody synthesis.
Caution: Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Storage Temperature:
Ambient |
MDL Number:
MFCD00081561 CAS Number: 9015-73-0 Merck Index: 14,2936 |
Specification Test Results
Form | Powder |
Free DEAE-Cl |
|
Loss on Drying |
|
Molecular Weight (viscosity calc.) | 500,000 ± 20% |
Total Nitrogen | 3.0 - 3.8% |
Optical purity | NMT 0.2 absorption at 375 nm |
pH (aqueous solution) | 5.0 to 7.0 |
Sulfated Ash |
|
Viscosity | 0.4 to 0.6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...