Diacerein

Supplier: Thermo Scientific Chemicals

J63775-03 J63775-06
CAAAJ63775-03EA 341.71 CAD
CAAAJ63775-03 CAAAJ63775-06
Diacerein
Diacerein

Interleukin-1 inhibitor


Powder


Applications: Interleukin-1 inhibitor

Formula: C₁₉H₁₂O₈
Melting Pt: 217…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00468030
CAS Number: 13739-02-1
EINECS: 237-310-2
Merck Index: 14,2960

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR