Gossypol ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,2'-bis(8-Formyl-1,6,7-trihydroxy-5-isopropyl-3-methylnaphthalene), 1,1',6,6',7,7'-Hexahydroxy-5,5'-diisopropyl-3,3'-dimethyl-[2,2'-binaphthalene]-8,8'-dicarbaldehyde, Thespesin

J63767-MC J63767-MD
CAAAJ63767-MCEA 461.9 CAD
CAAAJ63767-MC CAAAJ63767-MD
Gossypol ≥98%
Gossypol

Lipophilic agent derived from cottonseed


Powder


Applications: Lipophilic agent derived from cottonseed

Formula: C₃₀H₃₀O₈
Storage Temperature: Refrigerator
MDL Number: MFCD00017352
CAS Number: 303-45-7
Merck Index: 14,4528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR