Difloxacin hydrochloride

Supplier: Thermo Scientific Chemicals

J63754-06 J63754-14
CAAAJ63754-06EA 403.99 CAD
CAAAJ63754-06 CAAAJ63754-14
Difloxacin hydrochloride
Difloxacin hydrochloride

A quinolone antibacterial compound. Structurally related to norfloxacin


White powder


Applications: A quinolone antibacterial compound. Structurally related to norfloxacin

Formula: C₂₁H₁₉F₂N₃O₃·HCl
Storage Temperature: Ambient
MDL Number: MFCD03840489
CAS Number: 91296-86-5
Merck Index: 14,3141

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR