L(+)-Cysteine cell culture reagent
Supplier: Thermo Scientific Chemicals
|
Warning
|
J63745-14
J63745-22
CAAAJ63745-14EA
91.23
CAD
CAAAJ63745-14
CAAAJ63745-22
L(+)-Cysteine cell culture reagent
L-Cysteine
Powder
CAS No.: 52-90-4
Molecular Formula: C3H7NO2S
Formula Weight: 121.16
Physical Form: Powder
Sensitivity: Air Sensitive
Appearance: White
Melting Point: ca 220° dec.
Beilstein Reference: 1721408
Merck Reference: 14,2781
Optical Rotation: +7.6° (c=5 in 5N HCl)
MDL No.: MFCD00064306
Formula:
HSCH₂CH(NH₂)CO₂H Boiling Pt: 254 °C (1013 hPa) Melting Pt: ∼ 220 °C (dec.) Density: 1.523 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064306 CAS Number: 52-90-4 EINECS: 200-158-2 Merck Index: 14,2781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...