Potassium chloride 1 M aqueous solution

Supplier: Thermo Scientific Chemicals

Synonyms: Potassiumchloride

J63739AE J63739-AK
CAAAJ63739-AEEA 131.77 CAD
CAAAJ63739-AE CAAAJ63739-AK
Potassium chloride 1 M aqueous solution
Potassium chloride

Liquid


CAS No.: 7447-40-7 Molecular Formula: ClK Formula Weight: 74.55 Storage Temperature: 2°C to 8°C Physical Form: Liquid

Formula: KCl
MW: 74.55 g/mol
Boiling Pt: 1420
Melting Pt: 773
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8
Merck Index: 14,07621

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR