L-Cystine dihydrochloride 98% (by titrimetric analysis on dry substance)

Supplier: Thermo Scientific Chemicals

Synonyms: (2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid) dihydrochloride, (2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid) dihydrochloride, H-Cystine-OH·2HCl

J63717-30 J63717-A1
CAAAJ63717-30EA 391.22 CAD
CAAAJ63717-30 CAAAJ63717-A1
L-Cystine dihydrochloride 98% (by titrimetric analysis on dry substance)
L-Cystine dihydrochloride

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.


CAS No.: 30925-07-6 Molecular Formula: C6H12N2O4S2.2HCl Formula Weight: 313.22 Physical Form: Powder Appearance: White to pale yellow Melting Point: 228-232° MDL No.: MFCD00070399

Formula: C₆H₁₂N₂O₄S₂·2HCl
MW: 313.23 g/mol
Melting Pt: 228…232 °C
Storage Temperature: Ambient
MDL Number: MFCD00070399
CAS Number: 30925-07-6
EINECS: 250-391-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR