Carbenoxolone disodium salt ≥97%

Supplier: Thermo Scientific Chemicals
Warning

J63714-06 J63714-03
CAAAJ63714-06EA 352.45 CAD
CAAAJ63714-06 CAAAJ63714-03
Carbenoxolone disodium salt ≥97%
Carbenoxolone disodium salt

A triterpenoid chemopreventive agent


Solid


Applications: A triterpenoid chemopreventive agent

Formula: C₃₄H₄₈Na₂O₇
Storage Temperature: Ambient
MDL Number: MFCD00079043
CAS Number: 7421-40-1
EINECS: 231-044-0
Merck Index: 14,1793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR