L-Threonine cell culture reagent
Supplier: Thermo Scientific Chemicals
J63709-30
J63709-A1
CAAAJ63709-30EA
76.06
CAD
CAAAJ63709-30
CAAAJ63709-A1
L-Threonine cell culture reagent
L-Threonine
Powder
CAS No.: 72-19-5
Molecular Formula: C4H9NO3
Formula Weight: 119.12
Physical Form: Powder
Appearance: White
Melting Point: 256° dec.
Beilstein Reference: 1721646
Merck Reference: 14,9380
MDL No.: MFCD00064270
Formula:
C₄H₉NO₃ Melting Pt: 256 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00064270 CAS Number: 72-19-5 EINECS: 200-774-1 Merck Index: 14,9380 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...